Assay Details
Target Gene Details
Entrez Gene ID: | 221895 |
Gene Name: | JAZF zinc finger 1 |
Gene Aliases: |
TIP27, ZNF802 |
Location: |
Chr.7:27830573-28180818 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAZF1 | NM_175061.3 | 5 | 1355 | NP_778231.2 |
| XM_006715656.1 | 6 | 1231 | XP_006715719.1 | |
| XM_017011831.1 | 5 | 1123 | XP_016867320.1 | |
| XM_017011832.1 | 5 | 1055 | XP_016867321.1 | |
| AK000711.1 | 2 | 582 | ||
| AK091311.1 | 5 | 1280 | ||
| AL832491.1 | 3 | 933 | CAH10579.2 | |
| AL834234.1 | 4 | 1074 | CAD38912.1 | |
| AY372319.1 | 5 | 1133 | AAQ74874.1 | |
| BC042441.1 | 6 | 1143 | AAH42441.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5677 | Chr.7:27799885 - 27844886 on Build GRCh38 | Deletion |
|
| nsv1030685 | Chr.7:27804419 - 28137682 on Build GRCh38 | Loss |
|
| esv3891091 | Chr.7:27821143 - 28105945 on Build GRCh38 | Gain |
|
| esv2759520 | Chr.7:27753997 - 27933250 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map