Assay Details
Target Gene Details
Entrez Gene ID: | 3920 |
Gene Name: | lysosomal associated membrane protein 2 |
Gene Aliases: |
CD107b, LAMP-2, LAMPB, LGP110 |
Location: |
Chr.X:120426148-120469349 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMP2 | NM_001122606.1 | NP_001116078.1 | ||
| NM_002294.2 | NP_002285.1 | |||
| NM_013995.2 | 9 | 2199 | NP_054701.1 | |
| AK291090.1 | ||||
| AK295038.1 | ||||
| AY561849.1 | AAS67876.1 | |||
| BC002965.1 | 9 | 2154 | AAH02965.1 | |
| J04183.1 | AAA60383.1 | |||
| S79873.1 | 9 | 2156 | AAB35426.1 | |
| U36336.1 | 9 | 2156 | AAA91149.1 | |
| X77196.1 | CAA54416.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528786 | Chr.X:120380837 - 120471759 on Build GRCh38 | Gain |
|
| esv2758889 | Chr.X:120267570 - 120449094 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map