Assay Details
Target Gene Details
Entrez Gene ID: | 270 |
Gene Name: | adenosine monophosphate deaminase 1 |
Gene Aliases: |
MAD, MADA, MMDD |
Location: |
Chr.1:114673098-114695618 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMPD1 | NM_000036.2 | 7 | 976 | NP_000027.2 |
| NM_001172626.1 | 6 | 964 | NP_001166097.1 | |
| AK097077.1 | 6 | 964 | ||
| AK291349.1 | 7 | 880 | ||
| AK314252.1 | 7 | 886 | ||
| BC056678.1 | 7 | 905 | AAH56678.1 | |
| M60092.1 | 7 | 913 | AAA57281.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map