Assay Details
Target Gene Details
Entrez Gene ID: | 5888 |
Gene Name: | RAD51 recombinase |
Gene Aliases: |
BRCC5, FANCR, HRAD51, HsRad51, HsT16930, MRMV2, RAD51A, RECA |
Location: |
Chr.15:40694774-40732340 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51 | NM_001164269.1 | 10 | 1467 | NP_001157741.1 |
| NM_001164270.1 | 9 | 1497 | NP_001157742.1 | |
| NM_002875.4 | 10 | 1619 | NP_002866.2 | |
| NM_133487.3 | 10 | 1622 | NP_597994.3 | |
| XM_006720626.3 | 10 | 1670 | XP_006720689.1 | |
| XM_011521857.2 | 10 | 1369 | XP_011520159.1 | |
| XM_011521858.2 | 10 | 1416 | XP_011520160.1 | |
| XM_011521859.2 | 10 | 1553 | XP_011520161.1 | |
| XM_011521860.2 | 10 | 1464 | XP_011520162.1 | |
| XM_011521861.2 | 9 | 1247 | XP_011520163.1 | |
| XM_011521862.2 | 9 | 1469 | XP_011520164.1 | |
| AK131299.1 | 10 | 1467 | BAD18467.1 | |
| AK303089.1 | 6 | 1033 | ||
| BC001459.2 | 8 | 1285 | AAH01459.1 | |
| D13804.1 | 10 | 1531 | BAA02962.1 | |
| D14134.1 | 10 | 1552 | BAA03189.1 | |
| EU362635.1 | 9 | 1497 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527234 | Chr.15:40695527 - 40739488 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map