Assay Details
Target Gene Details
Entrez Gene ID: | 7252 |
Gene Name: | thyroid stimulating hormone beta |
Gene Aliases: |
TSH-B, TSH-BETA |
Location: |
Chr.1:115029824-115035935 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSHB | XM_011542065.2 | 2 | 676 | XP_011540367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26121 | Chr.1:115020681 - 115560070 on Build GRCh38 | Loss |
|
| nsv547570 | Chr.1:115029744 - 115066582 on Build GRCh38 | Loss |
|
| nsv528883 | Chr.1:114876400 - 115048377 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map