Assay Details
Target Gene Details
Entrez Gene ID: | 2739 |
Gene Name: | glyoxalase I |
Gene Aliases: |
GLOD1, GLYI, HEL-S-74 |
Location: |
Chr.6:38675925-38703176 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLO1 | NM_006708.2 | 6 | 1886 | NP_006699.2 |
| AB209801.1 | 6 | 1777 | BAD93038.1 | |
| AK129573.1 | 6 | 1819 | ||
| AK130312.1 | 6 | 1783 | ||
| AL833442.1 | 6 | 1836 | ||
| BC001741.1 | 6 | 1827 | AAH01741.1 | |
| BC011365.1 | 6 | 1840 | AAH11365.1 | |
| BC015934.1 | 6 | 1811 | AAH15934.1 | |
| BU954454.1 | 1 | 146 | ||
| D13315.1 | 6 | 1851 | BAA02572.1 | |
| FJ224325.1 | 6 | 1851 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428144 | Chr.6:38621193 - 38743309 on Build GRCh38 | Gain |
|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map