Assay Details
Target Gene Details
Entrez Gene ID: | 221895 |
Gene Name: | JAZF zinc finger 1 |
Gene Aliases: |
TIP27, ZNF802 |
Location: |
Chr.7:27830573-28180818 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAZF1 | NM_175061.3 | 1 | 315 | NP_778231.2 |
| XM_006715656.1 | 1 | 129 | XP_006715719.1 | |
| AK091311.1 | 1 | 240 | ||
| AY372319.1 | 1 | 93 | AAQ74874.1 | |
| BC042441.1 | 1 | 103 | AAH42441.1 | |
| DA864624.1 | 1 | 315 |
Target Gene Details
Entrez Gene ID: | 100128081 |
Gene Name: | JAZF1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:28180457-28241377 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAZF1-AS1 | NR_034097.1 | |||
| AK293540.1 | ||||
| DB481568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032041 | Chr.7:28149930 - 28183562 on Build GRCh38 | Gain |
|
| nsv950820 | Chr.7:28179682 - 28180781 on Build GRCh38 | Deletion |
|
| nsv830931 | Chr.7:28132064 - 28329752 on Build GRCh38 | Gain |
|
| nsv1137575 | Chr.7:28179681 - 28180781 on Build GRCh38 | Deletion |
|
| nsv1074083 | Chr.7:28179680 - 28180781 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map