Assay Details
Target Gene Details
Entrez Gene ID: | 101928881 |
Gene Name: | uncharacterized LOC101928881 |
Gene Aliases: |
- |
Location: |
Chr.2:233012614-233015885 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928881 | NR_136325.1 | 2 | 403 | |
| AI380278.1 | 2 | 253 |
Target Gene Details
Entrez Gene ID: | 25791 |
Gene Name: | neuronal guanine nucleotide exchange factor |
Gene Aliases: |
ARHGEF27, EPHEXIN |
Location: |
Chr.2:232878686-233013241 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NGEF | NM_019850.2 | 1 | 56 | NP_062824.2 |
| AK298315.1 | ||||
| AK300953.1 | 1 | 87 | ||
| BC031573.1 | AAH31573.1 | |||
| DB242442.1 | 1 | 56 | ||
| DC388448.1 | 1 | 87 |
More Information
Set Membership: |
|

Genomic Map