Assay Details
Target Gene Details
Entrez Gene ID: | 79101 |
Gene Name: | TATA-box binding protein associated factor, RNA polymerase I subunit D |
Gene Aliases: |
JOSD3, RAFI41, TAF(I)41, TAFI41 |
Location: |
Chr.11:93735929-93741537 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF1D | NM_024116.3 | NP_077021.1 | ||
| AF086435.1 | ||||
| AF275800.1 | AAG23815.1 | |||
| AK027016.1 | BAB15628.1 | |||
| CR457369.1 | CAG33650.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832232 | Chr.11:93668148 - 93816272 on Build GRCh38 | Gain |
|
| esv3627294 | Chr.11:93485306 - 93944317 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map