Assay Details
Target Gene Details
Entrez Gene ID: | 9293 |
Gene Name: | G protein-coupled receptor 52 |
Gene Aliases: |
- |
Location: |
Chr.1:174448074-174449545 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR52 | NM_005684.4 | 1 | 545 | NP_005675.3 |
| BC067456.1 | 1 | 507 | AAH67456.1 | |
| BC067457.1 | 1 | 507 | AAH67457.1 | |
| BC067458.1 | 1 | 507 | AAH67458.1 | |
| BC067459.1 | 1 | 507 | AAH67459.1 | |
| BC067460.1 | 1 | 507 | AAH67460.1 | |
| BC069460.1 | 1 | 507 | AAH69460.1 | |
| BC095532.1 | 1 | 546 | AAH95532.1 | |
| HQ995526.1 | 1 | 507 |
Target Gene Details
Entrez Gene ID: | 9910 |
Gene Name: | RAB GTPase activating protein 1 like |
Gene Aliases: |
HHL, TBC1D18 |
Location: |
Chr.1:174159414-174995308 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RABGAP1L | NM_014857.4 | NP_055672.3 | ||
| XM_005245680.1 | XP_005245737.1 | |||
| XM_005245681.2 | XP_005245738.1 | |||
| XM_006711693.1 | XP_006711756.1 | |||
| XM_011510223.2 | XP_011508525.1 | |||
| XM_017002994.1 | XP_016858483.1 | |||
| AB449904.1 | ||||
| AF279778.1 | ||||
| AK074409.1 | ||||
| AK095838.2 | BAC04635.1 | |||
| AY364260.1 | AAQ76819.1 | |||
| BC041888.1 | AAH41888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548194 | Chr.1:174164113 - 174500767 on Build GRCh38 | Loss |
|
| nsv470746 | Chr.1:174181712 - 174500767 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map