Assay Details
Target Gene Details
Entrez Gene ID: | 9873 |
Gene Name: | FCH and double SH3 domains 2 |
Gene Aliases: |
NWK, SH3MD3 |
Location: |
Chr.11:72836743-73142682 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCHSD2 | NM_014824.2 | 14 | 1615 | NP_055639.2 |
| XM_011545409.1 | 14 | 1497 | XP_011543711.1 | |
| XM_011545410.2 | 14 | 1430 | XP_011543712.1 | |
| XM_017018632.1 | 14 | 1674 | XP_016874121.1 | |
| AB018312.1 | 13 | 1465 | BAA34489.2 | |
| AK296376.1 | 14 | 1357 | ||
| AK297928.1 | 14 | 1499 | ||
| AL133567.1 | 9 | 983 | CAB63720.1 | |
| BC010394.1 | 14 | 1362 | AAH10394.1 | |
| BC017751.1 | 2 | 165 | AAH17751.1 |
More Information
Set Membership: |
|

Genomic Map