Assay Details
Target Gene Details
Entrez Gene ID: | 27013 |
Gene Name: | cyclin Pas1/PHO80 domain containing 1 |
Gene Aliases: |
C2orf24, CGI-57 |
Location: |
Chr.2:219171897-219178174 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNPPD1 | NM_001321389.1 | 9 | 1836 | NP_001308318.1 |
| NM_001321390.1 | 9 | 1481 | NP_001308319.1 | |
| NM_001321391.1 | 9 | 1515 | NP_001308320.1 | |
| NM_015680.5 | 8 | 1573 | NP_056495.3 | |
| NR_135628.1 | 8 | 1453 | ||
| NR_135629.1 | 7 | 1511 | ||
| AF070638.1 | 2 | 835 | AAC25393.1 | |
| AF151815.1 | 9 | 1740 | AAD34052.1 | |
| AK075274.1 | 8 | 1529 | ||
| AY007160.1 | 8 | 1546 | AAG02008.1 | |
| BC001393.2 | 8 | 1496 | AAH01393.1 | |
| BC012821.2 | 8 | 1545 | AAH12821.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508200 | Chr.2:219134744 - 219202175 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map