Assay Details
Target Gene Details
Entrez Gene ID: | 80834 |
Gene Name: | taste 1 receptor member 2 |
Gene Aliases: |
GPR71, T1R2, TR2 |
Location: |
Chr.1:18839599-18859661 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAS1R2 | NM_152232.2 | 1 | 115 | NP_689418.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs72953144] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map