Assay Details
Target Gene Details
Entrez Gene ID: | 389763 |
Gene Name: | SPATA31 subfamily D member 1 |
Gene Aliases: |
FAM75D1 |
Location: |
Chr.9:81987404-81995256 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA31D1 | NM_001001670.2 | 4 | 3456 | NP_001001670.1 |
| XM_017014710.1 | 4 | 3434 | XP_016870199.1 | |
| AK128831.1 | 4 | 3456 | BAC87631.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831639 | Chr.9:81886633 - 82036236 on Build GRCh38 | Loss |
|
| nsv821665 | Chr.9:80752718 - 82292739 on Build GRCh38 | Loss |
|
| nsv8537 | Chr.9:81987614 - 81996433 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map