Assay Details
Target Gene Details
Entrez Gene ID: | 2162 |
Gene Name: | coagulation factor XIII A chain |
Gene Aliases: |
F13A |
Location: |
Chr.6:6144078-6320691 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F13A1 | NM_000129.3 | 15 | 2666 | NP_000120.2 |
| AB208852.1 | 15 | 2618 | BAD92089.1 | |
| AK130666.1 | 8 | 1512 | ||
| BC027963.1 | 15 | 2624 | AAH27963.1 | |
| M14354.1 | 15 | 2627 | AAA52488.1 | |
| M14539.1 | 15 | 2630 | AAA52489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436891 | Chr.6:5693569 - 6189552 on Build GRCh38 | Insertion |
|
| dgv1709e212 | Chr.6:6132203 - 6147984 on Build GRCh38 | Loss |
|
| nsv1031283 | Chr.6:6084326 - 6159806 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map