Assay Details
Target Gene Details
Entrez Gene ID: | 1949 |
Gene Name: | ephrin B3 |
Gene Aliases: |
EFL6, EPLG8, LERK8 |
Location: |
Chr.17:7705202-7711375 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFNB3 | NM_001406.3 | 5 | 2131 | NP_001397.1 |
| AK027329.1 | 2 | 1254 | ||
| BC022499.1 | 4 | 1570 | AAH22499.1 | |
| BC042944.2 | 5 | 2131 | AAH42944.1 | |
| U66406.1 | 5 | 2347 | AAC51203.1 |
More Information
Set Membership: |
|

Genomic Map