Assay Details
Target Gene Details
Entrez Gene ID: | 23187 |
Gene Name: | pleckstrin homology like domain family B member 1 |
Gene Aliases: |
LL5A |
Location: |
Chr.11:118606296-118658038 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832283 | Chr.11:118566790 - 118786484 on Build GRCh38 | Loss |
|
| nsv527864 | Chr.11:118610716 - 118657714 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map