Assay Details
Target Gene Details
Entrez Gene ID: | 2005 |
Gene Name: | ELK4, ETS transcription factor |
Gene Aliases: |
SAP1 |
Location: |
Chr.1:205597774-205634077 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELK4 | BC040145.1 | AAH40145.1 |
Target Gene Details
Entrez Gene ID: | 148808 |
Gene Name: | major facilitator superfamily domain containing 4A |
Gene Aliases: |
MFSD4, UNQ3064 |
Location: |
Chr.1:205568984-205602918 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD4A | NM_181644.4 | 10 | 3147 | NP_857595.3 |
| XM_011509229.2 | 9 | 2886 | XP_011507531.1 | |
| AK090831.1 | 4 | 1874 | ||
| AK127321.1 | 2 | 2783 | ||
| AL713676.1 | 8 | 2453 | CAD28481.1 | |
| AY358107.1 | 8 | 2534 | AAQ88474.1 | |
| BC016377.1 | AAH16377.1 | |||
| BC036549.2 | 10 | 3097 | AAH36549.2 |
More Information
Set Membership: |
|

Genomic Map