Assay Details
Target Gene Details
Entrez Gene ID: | 5887 |
Gene Name: | RAD23 homolog B, nucleotide excision repair protein |
Gene Aliases: |
HHR23B, HR23B, P58 |
Location: |
Chr.9:107283236-107332194 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD23B | NM_002874.4 | 1 | 88 | NP_002865.1 |
| AK125226.1 | ||||
| AK223479.1 | BAD97199.1 | |||
| AK293532.1 | 1 | 80 | ||
| AL540969.3 | 1 | 58 | ||
| CA866470.1 | 1 | 71 | ||
| D21090.1 | BAA04652.1 | |||
| DC404422.1 | 1 | 109 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831682 | Chr.9:107174900 - 107313172 on Build GRCh38 | Gain |
|
| nsv508560 | Chr.9:107244209 - 107298258 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map