Assay Details
Target Gene Details
Entrez Gene ID: | 2244 |
Gene Name: | fibrinogen beta chain |
Gene Aliases: |
HEL-S-78p |
Location: |
Chr.4:154562980-154572763 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGB | NM_001184741.1 | 8 | 1574 | NP_001171670.1 |
| NM_005141.4 | 8 | 1751 | NP_005132.2 | |
| AI174804.1 | 1 | 408 | ||
| AK303788.1 | 9 | 1669 | ||
| BC107766.1 | 8 | 1720 | AAI07767.1 | |
| EU668333.1 | 8 | 1737 | ||
| J00129.1 | 8 | 1688 | AAA52429.1 |
More Information
Set Membership: |
|

Genomic Map