Assay Details
Target Gene Details
Entrez Gene ID: | 10269 |
Gene Name: | zinc metallopeptidase STE24 |
Gene Aliases: |
FACE-1, FACE1, HGPS, PRO1, STE24, Ste24p |
Location: |
Chr.1:40258050-40294184 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMPSTE24 | NM_005857.4 | NP_005848.2 | ||
| AB016068.1 | BAA33727.1 | |||
| AF064867.1 | AAC68866.1 | |||
| AK027874.1 | ||||
| AK074784.1 | ||||
| AK075007.1 | ||||
| BC037283.1 | AAH37283.1 | |||
| Y13834.1 | CAB46277.1 |
More Information
Set Membership: |
|

Genomic Map