Assay Details
Target Gene Details
Entrez Gene ID: | 159090 |
Gene Name: | family with sequence similarity 122B |
Gene Aliases: |
SPACIA2 |
Location: |
Chr.X:134769566-134797232 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM122B | NM_001166599.2 | 9 | 2407 | NP_001160071.1 |
| NM_001166600.2 | NP_001160072.1 | |||
| NM_001170756.1 | 10 | 1664 | NP_001164227.1 | |
| NM_001170757.1 | NP_001164228.1 | |||
| NM_145284.5 | 9 | 1610 | NP_660327.2 | |
| XM_006724730.2 | 10 | 2523 | XP_006724793.1 | |
| XM_006724732.2 | XP_006724795.1 | |||
| XM_006724733.3 | XP_006724796.1 | |||
| XM_011531282.1 | XP_011529584.1 | |||
| XM_011531283.1 | XP_011529585.1 | |||
| XM_011531284.1 | XP_011529586.1 | |||
| XM_011531285.1 | XP_011529587.1 | |||
| XM_011531286.1 | XP_011529588.1 | |||
| XM_011531287.1 | 10 | 2535 | XP_011529589.1 | |
| XM_011531288.1 | XP_011529590.1 | |||
| XM_011531289.1 | 10 | 2532 | XP_011529591.1 | |
| XM_011531290.1 | XP_011529592.1 | |||
| XM_011531291.1 | XP_011529593.1 | |||
| XM_011531292.1 | XP_011529594.1 | |||
| XM_011531293.1 | XP_011529595.1 | |||
| XM_011531294.1 | XP_011529596.1 | |||
| XM_017029306.1 | XP_016884795.1 | |||
| XM_017029307.1 | XP_016884796.1 | |||
| AB541015.1 | ||||
| AK026809.1 | ||||
| AK124650.1 | BAC85918.1 | |||
| AK124940.1 | BAC85999.1 | |||
| AK125991.1 | 10 | 1567 | BAC86380.1 | |
| AK297720.1 | ||||
| AL832218.1 | 9 | 1540 | ||
| BC110846.1 | AAI10847.1 | |||
| BX538218.1 | 10 | 1887 | CAD98073.1 |

Genomic Map