Assay Details
Target Gene Details
Entrez Gene ID: | 221150 |
Gene Name: | spindle and kinetochore associated complex subunit 3 |
Gene Aliases: |
C13orf3, RAMA1 |
Location: |
Chr.13:21153595-21176602 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKA3 | NM_001166017.1 | 8 | 1542 | NP_001159489.1 |
| NM_145061.5 | 9 | 1661 | NP_659498.4 | |
| XM_005266288.3 | 8 | 1447 | XP_005266345.1 | |
| XM_011534994.1 | 9 | 1637 | XP_011533296.1 | |
| AF361358.1 | 9 | 1603 | AAO13796.1 | |
| AK090584.1 | 8 | 1487 | BAC03484.1 | |
| BC048988.1 | 9 | 1632 | AAH48988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv315n67 | Chr.13:21153758 - 21155122 on Build GRCh38 | Gain |
|
| esv28665 | Chr.13:21153092 - 21155852 on Build GRCh38 | Gain |
|
| dgv3077n54 | Chr.13:21153703 - 21155087 on Build GRCh38 | Gain |
|
| nsv561256 | Chr.13:21154290 - 21155087 on Build GRCh38 | Loss |
|
| dgv960e59 | Chr.13:21153513 - 21155311 on Build GRCh38 | Duplication |
|
| nsv512307 | Chr.13:21154471 - 21157207 on Build GRCh38 | Loss |
|
| esv32897 | Chr.13:21154210 - 21155672 on Build GRCh38 | Loss |
|
| nsv1037082 | Chr.13:20932196 - 21204957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map