Assay Details
Target Gene Details
Entrez Gene ID: | 6924 |
Gene Name: | transcription elongation factor B subunit 3 |
Gene Aliases: |
EloA, SIII, SIII p110, TCEB3A |
Location: |
Chr.1:23743366-23762059 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEB3 | NM_003198.2 | 3 | 323 | NP_003189.2 |
| AF116729.1 | 3 | 172 | AAO15305.1 | |
| AK310229.1 | 3 | 309 | ||
| AK313079.1 | 3 | 195 | ||
| BC002883.2 | 3 | 207 | AAH02883.1 | |
| DB461774.1 | 3 | 361 | ||
| L47345.1 | 3 | 217 | AAA75492.1 |
More Information
Set Membership: |
|

Genomic Map