Assay Details
Target Gene Details
Entrez Gene ID: | 26999 |
Gene Name: | cytoplasmic FMR1 interacting protein 2 |
Gene Aliases: |
PIR121 |
Location: |
Chr.5:157266080-157395598 on Build GRCh38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYFIP2 | NM_001037333.2 | 31 | 3798 | NP_001032410.1 |
| NM_001291721.1 | 30 | 3720 | NP_001278650.1 | |
| NM_001291722.1 | 32 | 3873 | NP_001278651.1 | |
| NM_014376.3 | 31 | 3924 | NP_055191.2 | |
| XM_011534516.2 | 31 | 4064 | XP_011532818.1 | |
| XM_017009341.1 | 31 | 3913 | XP_016864830.1 | |
| XM_017009342.1 | 32 | 3835 | XP_016864831.1 | |
| AB032994.2 | 32 | 3813 | ||
| AF160973.1 | 31 | 3707 | AAD45723.1 | |
| AK223604.1 | 31 | 3751 | BAD97324.1 | |
| AK294220.1 | 26 | 3167 | ||
| AK295865.1 | 28 | 3347 | ||
| AK297401.1 | 29 | 3530 | ||
| AK303702.1 | 30 | 3673 | ||
| AL136549.1 | 31 | 3797 | CAB66484.1 | |
| AL161999.1 | 23 | 2758 | CAB82329.1 | |
| BC011762.1 | 31 | 3924 | AAH11762.1 | |
| L47738.1 | 20 | 2573 | AAA79022.1 |
Target Gene Details
Entrez Gene ID: | 102724404 |
Gene Name: | uncharacterized LOC102724404 |
Gene Aliases: |
- |
Location: |
Chr.5:157362615-157460100 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724404 | NR_136204.1 | |||
| NR_136205.1 | ||||
| BI755909.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830526 | Chr.5:157325531 - 157527933 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map