Assay Details
Target Gene Details
Entrez Gene ID: | 23023 |
Gene Name: | transmembrane and coiled-coil domain family 1 |
Gene Aliases: |
- |
Location: |
Chr.3:129647792-129893605 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entrez Gene ID: | 23023 |
Gene Name: | transmembrane and coiled-coil domain family 1 |
Gene Aliases: |
- |
Location: |
Chr.3:129647792-129893605 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |