Assay Details
Target Gene Details
Entrez Gene ID: | 56952 |
Gene Name: | phosphoribosyl transferase domain containing 1 |
Gene Aliases: |
HHGP |
Location: |
Chr.10:24848607-24952644 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRTFDC1 | NM_001282786.1 | 8 | 906 | NP_001269715.1 |
| NM_020200.6 | 9 | 983 | NP_064585.1 | |
| AB209792.1 | BAD93029.1 | |||
| AK021950.1 | 9 | 943 | BAB13944.1 | |
| AK222674.1 | 9 | 942 | BAD96394.1 | |
| AK294130.1 | 8 | 865 | ||
| BC008662.2 | 9 | 945 | AAH08662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819709 | Chr.10:24849002 - 24849673 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map