Assay Details
Target Gene Details
Entrez Gene ID: | 7166 |
Gene Name: | tryptophan hydroxylase 1 |
Gene Aliases: |
TPRH, TRPH |
Location: |
Chr.11:18020537-18040788 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPH1 | NM_004179.2 | 1 | 77 | NP_004170.1 |
| BC106739.1 | 1 | 51 | AAI06740.1 | |
| BC106740.2 | 1 | 51 | ||
| BP423529.1 | 2 | 146 | ||
| X52836.1 | 1 | 51 | CAA37018.1 |
More Information
Set Membership: |
|

Genomic Map