Assay Details
Target Gene Details
Entrez Gene ID: | 91894 |
Gene Name: | chromosome 11 open reading frame 52 |
Gene Aliases: |
- |
Location: |
Chr.11:111918877-111926871 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C11orf52 | NM_080659.2 | 4 | 399 | NP_542390.2 |
| AK057948.1 | 4 | 337 | BAB71620.1 | |
| BC008501.1 | 4 | 363 | AAH08501.1 | |
| BQ102454.1 | 4 | 399 |
Target Gene Details
Entrez Gene ID: | 100528019 |
Gene Name: | HSPB2-C11orf52 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:111912736-111926871 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPB2-C11orf52 | NR_037651.1 | 5 | 786 | |
| BC110872.1 | 5 | 781 | AAI10873.1 |

Genomic Map