Assay Details
Target Gene Details
Entrez Gene ID: | 89849 |
Gene Name: | autophagy related 16 like 2 |
Gene Aliases: |
ATG16B, WDR80 |
Location: |
Chr.11:72814406-72843740 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG16L2 | NM_001318766.1 | 1 | 111 | NP_001305695.1 |
| NM_033388.1 | 1 | 111 | NP_203746.1 | |
| XM_005274376.4 | 1 | 110 | XP_005274433.1 | |
| XM_006718732.2 | 1 | 110 | XP_006718795.1 | |
| XM_006718733.3 | 1 | 110 | XP_006718796.1 | |
| XM_011545332.1 | 1 | 110 | XP_011543634.1 | |
| XM_011545333.1 | 1 | 90 | XP_011543635.1 | |
| AK090597.1 | 1 | 102 | BAC03485.1 | |
| AK091151.1 | 1 | 90 | ||
| AK093017.1 | 1 | 84 | BAC04021.1 | |
| AK303270.1 | 1 | 92 | ||
| BC108672.1 | 1 | 69 | AAI08673.1 | |
| BC137489.1 | 1 | 209 | ||
| BC137490.1 | 1 | 209 | ||
| BC142718.1 | 1 | 98 | ||
| BC146660.1 | 1 | 98 | ||
| BQ929430.1 | 1 | 122 |
More Information
Set Membership: |
|

Genomic Map