Assay Details
Target Gene Details
Entrez Gene ID: | 3697 |
Gene Name: | inter-alpha-trypsin inhibitor heavy chain 1 |
Gene Aliases: |
H1P, IATIH, IGHEP1, ITI-HC1, ITIH, SHAP |
Location: |
Chr.3:52777586-52792068 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITIH1 | NM_002215.3 | NP_002206.2 | ||
| AK292750.1 | ||||
| AK298398.1 | ||||
| AK298423.1 | ||||
| AK314198.1 | ||||
| BC069464.1 | AAH69464.1 | |||
| BX496392.1 | ||||
| X63652.1 | CAA45188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508928 | Chr.3:52769883 - 52888575 on Build GRCh38 | Insertion |
|
| nsv1010761 | Chr.3:52770531 - 52817242 on Build GRCh38 | Gain |
|
| esv2762328 | Chr.3:52740868 - 52968134 on Build GRCh38 | Loss |
|
| nsv834698 | Chr.3:52715967 - 52894203 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map