Assay Details
Target Gene Details
Entrez Gene ID: | 79567 |
Gene Name: | family with sequence similarity 65 member A |
Gene Aliases: |
- |
Location: |
Chr.16:67528814-67546788 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM65A | NM_001193522.1 | NP_001180451.1 | ||
| NM_001193523.1 | NP_001180452.1 | |||
| NM_001193524.1 | NP_001180453.1 | |||
| NM_024519.3 | NP_078795.2 | |||
| XM_011523321.1 | XP_011521623.1 | |||
| XM_011523322.1 | XP_011521624.1 | |||
| XM_011523324.2 | XP_011521626.1 | |||
| XM_011523325.1 | XP_011521627.1 | |||
| XM_011523326.1 | XP_011521628.1 | |||
| XM_017023663.1 | XP_016879152.1 | |||
| XM_017023664.1 | XP_016879153.1 | |||
| AB067517.1 | BAB67823.1 | |||
| AK097087.1 | ||||
| AK127792.1 | BAC87139.1 | |||
| AK293748.1 | ||||
| AK295677.1 | ||||
| AK307467.1 | ||||
| CN401995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833266 | Chr.16:67408564 - 67571421 on Build GRCh38 | Loss |
|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv827711 | Chr.16:67536103 - 67583824 on Build GRCh38 | Gain |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map