Assay Details
Target Gene Details
Entrez Gene ID: | 64755 |
Gene Name: | chromosome 16 open reading frame 58 |
Gene Aliases: |
RUS |
Location: |
Chr.16:31489464-31508419 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C16orf58 | NM_022744.3 | NP_073581.2 | ||
| XM_017023570.1 | XP_016879059.1 | |||
| AK000679.1 | ||||
| AK023930.1 | BAB14730.1 | |||
| AK092580.1 | 1 | 231 | BAC03918.1 | |
| AK222913.1 | BAD96633.1 | |||
| AK296166.1 | ||||
| AK296491.1 | 5 | 1005 | ||
| AK304608.1 | ||||
| BC000822.1 | AAH00822.1 | |||
| BC009308.2 | AAH09308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060240 | Chr.16:31484552 - 31570129 on Build GRCh38 | Gain |
|
| nsv833194 | Chr.16:31346858 - 31517224 on Build GRCh38 | Loss |
|
| esv3570465 | Chr.16:31450067 - 31915765 on Build GRCh38 | Gain |
|

Genomic Map