Assay Details
Target Gene Details
Entrez Gene ID: | 51094 |
Gene Name: | adiponectin receptor 1 |
Gene Aliases: |
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A |
Location: |
Chr.1:202940825-202958572 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADIPOR1 | NM_001290553.1 | NP_001277482.1 | ||
| NM_001290557.1 | NP_001277486.1 | |||
| NM_001290629.1 | NP_001277558.1 | |||
| NM_015999.5 | NP_057083.2 | |||
| XM_017001407.1 | XP_016856896.1 | |||
| AF125179.1 | AAQ13552.1 | |||
| AF125180.1 | AAQ13553.1 | |||
| AF151803.1 | AAD34040.1 | |||
| AK001484.1 | ||||
| AK058114.1 | ||||
| AK124455.1 | ||||
| AK222503.1 | BAD96223.1 | |||
| AY424279.1 | AAR08367.1 | |||
| BC001594.2 | AAH01594.1 | |||
| BC010743.1 | AAH10743.1 |
More Information
Set Membership: |
|

Genomic Map