Assay Details
Target Gene Details
Entrez Gene ID: | 64220 |
Gene Name: | stimulated by retinoic acid 6 |
Gene Aliases: |
MCOPCB8, MCOPS9, PP14296 |
Location: |
Chr.15:74179466-74212267 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRA6 | NM_001142617.1 | 19 | 2288 | NP_001136089.1 |
| NM_001142618.1 | 19 | 2270 | NP_001136090.1 | |
| NM_001142619.1 | 19 | 2261 | NP_001136091.1 | |
| NM_001199040.1 | 19 | 2404 | NP_001185969.1 | |
| NM_001199041.1 | 19 | 2415 | NP_001185970.1 | |
| NM_001199042.1 | 19 | 2519 | NP_001185971.1 | |
| NM_022369.3 | 19 | 2224 | NP_071764.3 | |
| XM_011521883.1 | 19 | 2190 | XP_011520185.1 | |
| XM_011521884.1 | 19 | 2331 | XP_011520186.1 | |
| XM_017022478.1 | 19 | 2139 | XP_016877967.1 | |
| XM_017022479.1 | 19 | 2230 | XP_016877968.1 | |
| XM_017022480.1 | 19 | 2254 | XP_016877969.1 | |
| AF352728.1 | 19 | 2090 | AAK30289.1 | |
| AF352729.1 | 19 | 2200 | AAK30290.1 | |
| AF370419.2 | 19 | 2196 | AAQ15255.2 | |
| AK022603.1 | 15 | 1774 | BAB14122.1 | |
| AK056125.1 | 19 | 2285 | ||
| AK091152.1 | 19 | 2314 | ||
| AK092227.1 | 14 | 1751 | ||
| AK291966.1 | ||||
| AK298921.1 | 19 | 2159 | ||
| AK298928.1 | ||||
| AK299191.1 | 19 | 2404 | ||
| AK302932.1 | 19 | 2415 | ||
| AK309676.1 | 19 | 2499 | ||
| AY358748.1 | 19 | 2200 | AAQ89108.1 | |
| AY359089.1 | 19 | 2090 | AAQ89447.1 | |
| BC025256.1 | 19 | 2196 | AAH25256.1 | |
| BX537413.1 | 19 | 2166 | CAD97655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss |
|
| nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain |
|
| nsv833054 | Chr.15:74165302 - 74332942 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map