Assay Details
Target Gene Details
Entrez Gene ID: | 63891 |
Gene Name: | ring finger protein 123 |
Gene Aliases: |
FP1477, KPC1 |
Location: |
Chr.3:49688753-49721529 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF123 | NM_022064.4 | 23 | 2190 | NP_071347.2 |
| NR_135218.1 | 23 | 2190 | ||
| XM_006713285.1 | 23 | 2348 | XP_006713348.1 | |
| XM_011533995.1 | 23 | 2125 | XP_011532297.1 | |
| XM_017007018.1 | 23 | 2152 | XP_016862507.1 | |
| AF370367.1 | 14 | 1344 | AAQ15203.1 | |
| AK022627.1 | BAB14139.1 | |||
| AK026968.1 | 5 | 444 | BAB15607.1 | |
| AK090632.1 | 13 | 1232 | ||
| AK096353.1 | 23 | 2104 | ||
| AK126883.1 | 9 | 739 | ||
| AK308130.1 | ||||
| AK308607.1 | 20 | 1868 | ||
| AL832034.1 | 23 | 2152 | ||
| AL832138.1 | 22 | 2354 | ||
| AL832344.1 | 23 | 2129 | ||
| AY744152.1 | 22 | 2073 | AAU93470.1 | |
| BC041145.1 | 8 | 723 | AAH41145.1 | |
| BC057392.1 | 10 | 880 | AAH57392.2 | |
| BC088801.1 | 5 | 451 | AAH88801.1 | |
| BC130632.1 | 23 | 2131 | ||
| CR627051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map