Assay Details
Target Gene Details
Entrez Gene ID: | 11227 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 5 |
Gene Aliases: |
GALNAC-T5, GALNACT5 |
Location: |
Chr.2:157257548-157318491 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT5 | NM_014568.1 | 1 | 434 | NP_055383.1 |
| XM_017003237.1 | 1 | 714 | XP_016858726.1 | |
| XM_017003238.1 | 1 | 714 | XP_016858727.1 | |
| AK292154.1 | 1 | 395 | ||
| AY277591.1 | 1 | 434 | AAP34404.1 | |
| BC142676.1 | 1 | 664 | ||
| BC142703.1 | 1 | 536 |
More Information
Set Membership: |
|

Genomic Map