Assay Details
Target Gene Details
Entrez Gene ID: | 146850 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 6 |
Gene Aliases: |
C17orf38, HsT41028, p84 PIKAP, p87(PIKAP), p87PIKAP |
Location: |
Chr.17:8802723-8870003 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3R6 | NM_001010855.3 | 13 | 1718 | NP_001010855.1 |
| NM_001290211.1 | 13 | 2086 | NP_001277140.1 | |
| NR_110865.1 | 12 | 1813 | ||
| XM_011523677.2 | 14 | 1728 | XP_011521979.1 | |
| XM_011523678.2 | 14 | 1715 | XP_011521980.1 | |
| XM_011523680.2 | XP_011521982.1 | |||
| XM_011523681.2 | 13 | 1979 | XP_011521983.1 | |
| XM_011523683.2 | 11 | 1416 | XP_011521985.1 | |
| XM_011523686.2 | 5 | 865 | XP_011521988.1 | |
| XM_017024228.1 | 14 | 1730 | XP_016879717.1 | |
| XM_017024229.1 | 14 | 1730 | XP_016879718.1 | |
| XM_017024230.1 | XP_016879719.1 | |||
| XM_017024231.1 | 10 | 1907 | XP_016879720.1 | |
| AK091819.1 | 12 | 1815 | ||
| AY753192.1 | 11 | 1478 | AAV30085.1 | |
| BC125169.1 | 8 | 1122 |
More Information
Set Membership: |
|

Genomic Map