Assay Details
Target Gene Details
Entrez Gene ID: | 55088 |
Gene Name: | coiled-coil domain containing 186 |
Gene Aliases: |
C10orf118 |
Location: |
Chr.10:114120862-114174312 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC186 | NM_001321829.1 | NP_001308758.1 | ||
| NM_018017.3 | NP_060487.2 | |||
| NM_153249.1 | NP_694981.1 | |||
| NR_135815.1 | ||||
| XM_011539915.2 | XP_011538217.1 | |||
| XM_011539916.2 | 1 | 106 | XP_011538218.1 | |
| AI143965.1 | 1 | 400 | ||
| AK092620.1 | BAC03926.1 | |||
| AK307517.1 | ||||
| BC020842.1 | AAH20842.1 | |||
| BC070368.1 | AAH70368.1 | |||
| BC107430.1 | AAI07431.1 | |||
| BX537964.1 | CAD97928.1 | |||
| BX643041.1 |
Target Gene Details
Entrez Gene ID: | 100302224 |
Gene Name: | microRNA 2110 |
Gene Aliases: |
hsa-mir-2110, mir-2110 |
Location: |
Chr.10:114174105-114174179 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR2110 | NR_031747.1 |

Genomic Map