Assay Details
Target Gene Details
Entrez Gene ID: | 51347 |
Gene Name: | TAO kinase 3 |
Gene Aliases: |
DPK, JIK, MAP3K18 |
Location: |
Chr.12:118149801-118372950 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map