Assay Details
Target Gene Details
Entrez Gene ID: | 80223 |
Gene Name: | RAB11 family interacting protein 1 |
Gene Aliases: |
NOEL1A, RCP, rab11-FIP1 |
Location: |
Chr.8:37858947-37899516 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB11FIP1 | NM_001002814.2 | 6 | 3740 | NP_001002814.2 |
| NM_025151.4 | 5 | 1838 | NP_079427.4 | |
| AF368294.1 | 5 | 1808 | AAM09571.1 | |
| AK026177.1 | 2 | 79 | ||
| AK026275.1 | BAB15424.1 | |||
| AK092296.1 | 4 | 1146 | ||
| AK122583.1 | 3 | 1879 | BAC56924.1 | |
| AK124422.1 | 1 | 642 | ||
| AK226111.1 | 6 | 3430 | ||
| AK291781.1 | 5 | 1835 | ||
| AL832155.1 | 3 | 1321 | ||
| AL833169.1 | 3 | 1149 | ||
| AY280966.1 | 3 | 2161 | AAQ18784.1 | |
| AY280968.1 | 6 | 3697 | AAQ18786.1 | |
| AY280969.1 | 2 | 2910 | AAQ18787.1 | |
| BC077720.1 | 5 | 1808 | AAH77720.1 | |
| DQ083995.1 | 5 | 1562 | AAY86040.1 | |
| DQ236342.1 | 5 | 1808 | ABB43161.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509261 | Chr.8:37767151 - 37881434 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map