Assay Details
Target Gene Details
Entrez Gene ID: | 1999 |
Gene Name: | E74 like ETS transcription factor 3 |
Gene Aliases: |
EPR-1, ERT, ESE-1, ESX |
Location: |
Chr.1:202010519-202017188 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELF3 | NM_001114309.1 | NP_001107781.1 | ||
| NM_004433.4 | NP_004424.3 | |||
| AF016295.1 | AAB96586.1 | |||
| AF017307.1 | AAB67238.1 | |||
| AF517841.1 | AAM70481.1 | |||
| AK225446.1 | ||||
| AK310814.1 | ||||
| AK312273.1 | ||||
| BC003569.1 | AAH03569.1 | |||
| BX537368.1 | CAD97611.1 | |||
| CR457106.1 | CAG33387.1 | |||
| U66894.1 | AAB58075.1 | |||
| U73843.1 | AAB65823.1 | |||
| U73844.1 | AAB65824.1 | |||
| U97156.1 | AAC51884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516812 | Chr.1:202008699 - 202017290 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map