Assay Details
Target Gene Details
Entrez Gene ID: | 285193 |
Gene Name: | dual specificity phosphatase 28 |
Gene Aliases: |
DUSP26, VHP |
Location: |
Chr.2:240560054-240564014 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP28 | NM_001033575.1 | 3 | 1338 | NP_001028747.1 |
| BC036198.1 | 2 | 3244 | AAH36198.1 | |
| BQ777661.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759119 | Chr.2:240425704 - 240649454 on Build GRCh38 | Gain |
|
| esv3594802 | Chr.2:240487835 - 240574802 on Build GRCh38 | Gain |
|
| esv3575343 | Chr.2:240521892 - 240570726 on Build GRCh38 | Gain |
|
| nsv10226 | Chr.2:240482405 - 240575049 on Build GRCh38 | Gain |
|
| nsv999050 | Chr.2:240525384 - 240570295 on Build GRCh38 | Gain |
|
| nsv834586 | Chr.2:240475181 - 240622625 on Build GRCh38 | Loss |
|
| nsv1001262 | Chr.2:240482501 - 240671892 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map