Assay Details
Target Gene Details
Entrez Gene ID: | 79077 |
Gene Name: | dCTP pyrophosphatase 1 |
Gene Aliases: |
CDA03, RS21C6, XTP3TPA |
Location: |
Chr.16:30423698-30430052 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCTPP1 | NM_024096.1 | 3 | 748 | NP_077001.1 |
| NR_134470.1 | 3 | 773 | ||
| NR_134471.1 | 3 | 787 | ||
| AF210430.1 | 3 | 721 | AAK48422.1 | |
| AF212242.1 | 3 | 719 | AAK14927.1 | |
| AK024843.1 | 3 | 716 | BAB15025.1 | |
| AY453409.1 | 3 | 748 | AAR26724.1 | |
| BC001344.1 | 3 | 748 | AAH01344.1 | |
| BE897551.1 |
More Information
Set Membership: |
|

Genomic Map