Assay Details
Target Gene Details
Entrez Gene ID: | 29997 |
Gene Name: | glioma tumor suppressor candidate region gene 2 |
Gene Aliases: |
PICT-1, PICT1 |
Location: |
Chr.19:47745536-47757066 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLTSCR2 | NM_015710.4 | NP_056525.2 | ||
| AF182076.1 | AAF62873.1 | |||
| AF296124.1 | AAG30413.1 | |||
| AK024486.1 | BAB15776.1 | |||
| AK225915.1 | ||||
| AK297877.1 | ||||
| AK301117.1 | ||||
| AK309788.1 | ||||
| AL359335.1 | CAB94786.1 | |||
| AL359336.1 | CAB94787.1 | |||
| AY535000.1 | AAS46028.1 | |||
| BC004229.2 | AAH04229.2 | |||
| BC006311.2 | AAH06311.1 | |||
| BC007248.1 | AAH07248.1 | |||
| BC010095.2 | AAH10095.1 | |||
| BC014009.2 | AAH14009.2 | |||
| CX866974.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644559 | Chr.19:47747204 - 47907847 on Build GRCh38 | Gain |
|
| nsv1058351 | Chr.19:47730870 - 47779238 on Build GRCh38 | Gain |
|
| nsv1059189 | Chr.19:47695560 - 47768115 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map