Assay Details
Target Gene Details
Entrez Gene ID: | 90326 |
Gene Name: | THAP domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.1:6624645-6635586 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THAP3 | NM_001195752.1 | NP_001182681.1 | ||
| NM_001195753.1 | NP_001182682.1 | |||
| NM_138350.3 | NP_612359.2 | |||
| XM_005263532.3 | XP_005263589.1 | |||
| XM_011542400.2 | XP_011540702.1 | |||
| XM_011542401.2 | XP_011540703.1 | |||
| XM_017002761.1 | XP_016858250.1 | |||
| AK096217.1 | BAC04727.1 | |||
| BC005114.1 | AAH05114.1 | |||
| BC092427.1 | AAH92427.1 |
More Information
Set Membership: |
|

Genomic Map