Assay Details
Target Gene Details
Entrez Gene ID: | 1489 |
Gene Name: | cardiotrophin 1 |
Gene Aliases: |
CT-1, CT1 |
Location: |
Chr.16:30895824-30903560 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTF1 | NM_001142544.1 | 3 | 1537 | NP_001136016.1 |
| NM_001330.3 | 3 | 1540 | NP_001321.1 | |
| XM_011545759.2 | 3 | 1649 | XP_011544061.1 | |
| XM_011545760.2 | 3 | 1607 | XP_011544062.1 | |
| BC036787.1 | 3 | 1506 | AAH36787.1 | |
| BC064416.1 | 3 | 1540 | AAH64416.1 | |
| U43030.1 | AAA85229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457483 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
| dgv35n68 | Chr.16:30833730 - 31036730 on Build GRCh38 | Loss |
|
| nsv953802 | Chr.16:30876680 - 30999279 on Build GRCh38 | Deletion |
|
| nsv833189 | Chr.16:30808387 - 30949474 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map