Assay Details
Target Gene Details
Entrez Gene ID: | 60677 |
Gene Name: | CUGBP, Elav-like family member 6 |
Gene Aliases: |
BRUNOL6 |
Location: |
Chr.15:72284727-72320184 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELF6 | NM_001172684.1 | 12 | 2290 | NP_001166155.1 |
| NM_001172685.1 | 11 | 2063 | NP_001166156.1 | |
| NM_052840.4 | 13 | 2609 | NP_443072.3 | |
| AK124858.1 | 12 | 2163 | ||
| AK131098.1 | BAC85148.1 | |||
| BC030835.1 | 13 | 2312 | AAH30835.1 | |
| BU732838.1 | 1 | 500 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1601 | Chr.15:72257544 - 72292835 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map