Assay Details
Target Gene Details
Entrez Gene ID: | 5601 |
Gene Name: | mitogen-activated protein kinase 9 |
Gene Aliases: |
JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK |
Location: |
Chr.5:180233594-180292071 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759398 | Chr.5:180107477 - 180302944 on Build GRCh38 | Gain |
|
| nsv1033706 | Chr.5:179975769 - 180379356 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map