Assay Details
Target Gene Details
Entrez Gene ID: | 57190 |
Gene Name: | selenoprotein N, 1 |
Gene Aliases: |
CFTD, MDRS1, RSMD1, RSS, SELN |
Location: |
Chr.1:25800176-25818222 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPN1 | NM_020451.2 | 13 | 3249 | NP_065184.2 |
| NM_206926.1 | 12 | 3147 | NP_996809.1 | |
| AF166125.1 | 11 | 2838 | AAF21430.1 | |
| AJ306399.1 | 13 | 3249 | CAC83791.1 | |
| AK172860.1 | 1 | 1566 | ||
| AL110205.1 | 1 | 988 | ||
| BC015638.2 | 4 | 1914 | AAH15638.1 | |
| BC042154.2 | 4 | 1914 | AAH42154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7490 | Chr.1:25797857 - 25843537 on Build GRCh38 | Deletion |
|
| nsv460884 | Chr.1:25778482 - 25891593 on Build GRCh38 | Loss |
|
| nsv834491 | Chr.1:25726411 - 25914117 on Build GRCh38 | Loss |
|
| nsv950337 | Chr.1:25800110 - 25819709 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map